Assay Details
| Gene Symbol: SEC14L6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005568097.2 | XP_005568154.1 | 6 - 7 | 585 | 100 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

SEC14L6| Gene Symbol: SEC14L6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005568097.2 | XP_005568154.1 | 6 - 7 | 585 | 100 | |