Assay Details
| Gene Symbol: DNAJC15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005585746.2 | XP_005585803.1 | 4 - 5 | 790 | 68 | |
| XM_015439215.1 | XP_015294701.1 | 4 - 5 | 367 | 68 | ||
| GenBank mRNA | AB174473.1 | - | 4 - 5 | 318 | 68 | |
More Information
Set Membership: |
|

DNAJC15
Genomic Map