Assay Details
| Gene Symbol: AADACL3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005544757.2 | XP_005544814.1 | 3 - 4 | 1008 | 77 | |
| XM_015442556.1 | XP_015298042.1 | 4 - 5 | 791 | 77 | ||
More Information
Set Membership: |
|

AADACL3
Genomic Map