Assay Details
| Gene Symbol: ADA2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005568054.2 | XP_005568111.1 | 8 - 9 | 1519 | 64 | |
| XM_005568055.2 | XP_005568112.1 | 8 - 9 | 1263 | 64 | ||
| XM_005568056.2 | XP_005568113.1 | 6 - 7 | 748 | 64 | ||
More Information
Set Membership: |
|

ADA2
Genomic Map