Assay Details
| Gene Symbol: LOC102122509 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015430374.1 | XP_015285860.1 | 4 - 5 | 1687 | 178 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC102122509| Gene Symbol: LOC102122509 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015430374.1 | XP_015285860.1 | 4 - 5 | 1687 | 178 | |
Set Membership: |
|