Assay Details
| Gene Symbol: CHMP3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005575367.2 | XP_005575424.1 | 5 - 6 | 985 | 101 | |
| XM_015433455.1 | XP_015288941.1 | 5 - 6 | 749 | 101 | ||
| GenBank mRNA | AB169670.1 | - | 5 - 6 | 658 | 101 | |
More Information
Set Membership: |
|

CHMP3
Genomic Map