Assay Details
| Gene Symbol: PPP1R15A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005589819.2 | XP_005589876.1 | 2 - 3 | 2054 | 89 | |
| XM_005589820.2 | XP_005589877.1 | 3 - 4 | 1925 | 89 | ||
| GenBank mRNA | AB170462.1 | - | 3 - 4 | 1914 | 89 | |
More Information
Set Membership: |
|

PPP1R15A
Genomic Map