Assay Details
| Gene Symbol: ZNF213 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005591049.2 | XP_005591106.1 | 5 - 6 | 1364 | 76 | |
| XM_015442668.1 | XP_015298154.1 | 4 - 5 | 1013 | 76 | ||
More Information
Set Membership: |
|

ZNF213
Genomic Map