Assay Details
| Gene Symbol: NUPR1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005591645.2 | XP_005591702.1 | 2 - 3 | 732 | 55 | |
| XM_005591646.2 | XP_005591703.1 | 2 - 3 | 652 | 55 | ||
More Information
Set Membership: |
|

NUPR1
Genomic Map