Assay Details
| Gene Symbol: CD24 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005552236.2 | XP_005552293.1 | 2 - 3 | 1057 | 58 | |
| XM_015449017.1 | XP_015304503.1 | 2 - 3 | 821 | 58 | ||
More Information
Set Membership: |
|

CD24
Genomic Map