Assay Details
| Gene Symbol: RAMP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005574769.2 | XP_005574826.1 | 2 - 3 | 316 | 86 | |
| XM_015433033.1 | XP_015288519.1 | 2 - 3 | 491 | 86 | ||
More Information
Set Membership: |
|

RAMP1
Genomic Map