Assay Details
| Gene Symbol: DTX3L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005547980.2 | XP_005548037.1 | 2 - 3 | 541 | 83 | |
| XM_005547981.2 | XP_005548038.1 | 2 - 3 | 370 | 83 | ||
More Information
Set Membership: |
|

DTX3L
Genomic Map