Assay Details
| Gene Symbol: LOC102146680 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005595395.2 | XP_005595452.1 | 1360 | 79 | ||
| XM_015444500.1 | XP_015299986.1 | 1922 | 79 | |||
| XM_015444501.1 | XP_015299987.1 | 814 | 79 | |||
More Information
Set Membership: |
|

LOC102146680
Genomic Map