Assay Details
| Gene Symbol: SHISAL2A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005543354.2 | XP_005543411.1 | 1 - 2 | 364 | 75 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

SHISAL2A| Gene Symbol: SHISAL2A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005543354.2 | XP_005543411.1 | 1 - 2 | 364 | 75 | |
Set Membership: |
|