Assay Details
| Gene Symbol: SHISAL2A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005543354.2 | XP_005543411.1 | 2 - 3 | 503 | 73 | |
| XM_015435803.1 | XP_015291289.1 | 2 - 3 | 1656 | 73 | ||
More Information
Set Membership: |
|

SHISAL2A
Genomic Map