Assay Details
| Gene Symbol: ZFP69B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005543833.2 | XP_005543890.1 | 3 - 4 | 625 | 73 | |
| XM_005543834.2 | XP_005543891.1 | 3 - 4 | 626 | 73 | ||
More Information
Set Membership: |
|

ZFP69B
Genomic Map