Assay Details
| Gene Symbol: ZFP69B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005543833.2 | XP_005543890.1 | 5 - 6 | 847 | 99 | |
| XM_005543834.2 | XP_005543891.1 | 5 - 6 | 848 | 99 | ||
| XM_015437781.1 | XP_015293267.1 | 4 - 5 | 761 | 99 | ||
More Information
Set Membership: |
|

ZFP69B
Genomic Map