Assay Details
| Gene Symbol: PSAPL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005554416.2 | XP_005554473.1 | 1 | 1352 | 86 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

PSAPL1| Gene Symbol: PSAPL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005554416.2 | XP_005554473.1 | 1 | 1352 | 86 | |
Set Membership: |
|