Assay Details
| Gene Symbol: GCNT4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005557163.2 | XP_005557220.1 | 2 - 3 | 1409 | 88 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

GCNT4| Gene Symbol: GCNT4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005557163.2 | XP_005557220.1 | 2 - 3 | 1409 | 88 | |
Set Membership: |
|