Assay Details
| Gene Symbol: NOXA1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005580295.2 | XP_005580352.1 | 8 - 9 | 875 | 98 | |
| XM_005580298.2 | XP_005580355.1 | 8 - 9 | 875 | 98 | ||
| XM_015436125.1 | XP_015291611.1 | 8 - 9 | 875 | 98 | ||
| XM_015436126.1 | XP_015291612.1 | 8 - 9 | 950 | 98 | ||
More Information
Set Membership: |
|

NOXA1
Genomic Map