Assay Details
| Gene Symbol: CFHR5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005540303.2 | XP_005540360.1 | 7 - 8 | 1254 | 66 | |
| XM_005540304.2 | XP_005540361.1 | 7 - 8 | 1271 | 66 | ||
More Information
Set Membership: |
|

CFHR5
Genomic Map