Assay Details
| Gene Symbol: LOC102123198 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001319474.1 | NP_001306403.1 | 14 - 15 | 1552 | 111 | |
| XM_015454704.1 | XP_015310190.1 | 14 - 15 | 1586 | 111 | ||
| GenBank mRNA | X76637.1 | - | 1552 | 111 | ||
More Information
Set Membership: |
|

LOC102123198
Genomic Map