Assay Details
| Gene Symbol: LOC102123198 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001319474.1 | NP_001306403.1 | 20 - 21 | 2227 | 100 | |
| GenBank mRNA | X76637.1 | - | 2227 | 100 | ||
More Information
Set Membership: |
|

LOC102123198
Genomic Map