Assay Details
| Gene Symbol: PATE1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005580091.2 | XP_005580148.1 | 4 - 5 | 264 | 76 | |
| XM_015436058.1 | XP_015291544.1 | 4 - 5 | 260 | 76 | ||
More Information
Set Membership: |
|

PATE1
Genomic Map