Assay Details
| Gene Symbol: MRPS2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005580467.2 | XP_005580524.1 | 3 - 4 | 403 | 65 | |
| XM_015436226.1 | XP_015291712.1 | 2 - 3 | 252 | 65 | ||
More Information
Set Membership: |
|

MRPS2
Genomic Map