Assay Details
| Gene Symbol: KLK15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005590042.2 | XP_005590099.1 | 3 - 4 | 1071 | 59 | |
| XM_005590045.2 | XP_005590102.1 | 3 - 4 | 1071 | 59 | ||
More Information
Set Membership: |
|

KLK15
Genomic Map