Assay Details
| Gene Symbol: ZC3H12D | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005552090.2 | XP_005552147.1 | 5 - 6 | 1583 | 67 | |
| XM_005552092.2 | XP_005552149.1 | 5 - 6 | 1535 | 67 | ||
| XM_015448948.1 | XP_015304434.1 | 5 - 6 | 1101 | 67 | ||
More Information
Set Membership: |
|

ZC3H12D
Genomic Map