Assay Details
| Gene Symbol: LOC101865550 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005556537.2 | XP_005556594.1 | 11 - 12 | 2900 | 93 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC101865550| Gene Symbol: LOC101865550 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005556537.2 | XP_005556594.1 | 11 - 12 | 2900 | 93 | |