Assay Details
| Gene Symbol: LOC101865550 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005556537.2 | XP_005556594.1 | 14 - 15 | 3061 | 99 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC101865550| Gene Symbol: LOC101865550 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005556537.2 | XP_005556594.1 | 14 - 15 | 3061 | 99 | |