Assay Details
| Gene Symbol: LOC102139567 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005584590.1 | XP_005584647.1 | 1 | 316 | 86 | |
| XM_015438646.1 | XP_015294132.1 | 1 | 316 | 86 | ||
| XM_015438647.1 | XP_015294133.1 | 1 | 316 | 86 | ||
| XM_015438649.1 | XP_015294135.1 | 1 | 316 | 86 | ||
| XM_015438650.1 | XP_015294136.1 | 1 | 316 | 86 | ||
| XM_015438651.1 | XP_015294137.1 | 1 | 316 | 86 | ||
| XM_015438652.1 | XP_015294138.1 | 1 | 316 | 86 | ||
| XM_015438653.1 | XP_015294139.1 | 1 | 316 | 86 | ||
| XM_015438655.1 | XP_015294141.1 | 1 | 316 | 86 | ||
| XM_015438656.1 | XP_015294142.1 | 1 | 316 | 86 | ||
More Information
Set Membership: |
|

LOC102139567
Genomic Map