Assay Details
| Gene Symbol: LOC102139613 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005590832.2 | XP_005590889.1 | 4 | 937 | 63 | |
| GenBank mRNA | EU341823.1 | - | 4 | 615 | 63 | |
More Information
Set Membership: |
|

LOC102139613
Genomic Map