Assay Details
| Gene Symbol: MTERF2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005572122.2 | XP_005572179.1 | 3 | 822 | 78 | |
| XM_005572123.2 | XP_005572180.1 | 2 | 698 | 78 | ||
| XM_015431487.1 | XP_015286973.1 | 3 | 832 | 78 | ||
| GenBank mRNA | AB174214.1 | - | 1 | 523 | 78 | |
More Information
Set Membership: |
|

MTERF2
Genomic Map