Assay Details
| Gene Symbol: GIMAP4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001283821.1 | NP_001270750.1 | 3 - 4 | 124 | 87 | |
| XM_015447315.1 | XP_015302801.1 | 2 - 3 | 87 | |||
| XM_015447316.1 | XP_015302802.1 | 2 - 3 | 87 | |||
| GenBank mRNA | AB174555.1 | - | 3 - 4 | 124 | 87 | |
More Information
Set Membership: |
|

GIMAP4
Genomic Map