Assay Details
| Gene Symbol: GIMAP4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001283821.1 | NP_001270750.1 | 4 - 5 | 856 | 117 | |
| GenBank mRNA | AB174555.1 | - | 4 - 5 | 856 | 117 | |
More Information
Set Membership: |
|

GIMAP4
Genomic Map