Assay Details
| Gene Symbol: INS | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001284919.1 | NP_001271848.1 | 2 - 3 | 249 | 103 | |
| XM_015434180.1 | XP_015289666.1 | 2 - 3 | 243 | 103 | ||
| GenBank mRNA | J00336.1 | - | 2 - 3 | 249 | 103 | |
More Information
Set Membership: |
|

INS
Genomic Map