Assay Details
| Gene Symbol: CCR9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001285292.1 | NP_001272221.1 | 1 | 1018 | 125 | |
| XM_015445355.1 | XP_015300841.1 | 3 | 1632 | 125 | ||
| XM_015445356.1 | XP_015300842.1 | 3 | 1872 | 125 | ||
| GenBank mRNA | DQ182547.1 | - | 1 | 1018 | 125 | |
More Information
Set Membership: |
|

CCR9
Genomic Map