Assay Details
| Gene Symbol: RAB9A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001283761.1 | NP_001270690.1 | 2 - 3 | 221 | 120 | |
| GenBank mRNA | AB172492.1 | - | 2 - 3 | 221 | 120 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001283761.1 transcripts.
Set Membership: |
|

RAB9A
Genomic Map