Assay Details
| Gene Symbol: GPR34 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005593329.2 | XP_005593386.1 | 3 | 1848 | 91 | |
| XM_005593330.2 | XP_005593387.1 | 3 | 1845 | 91 | ||
| XM_005593331.2 | XP_005593388.1 | 4 | 1866 | 91 | ||
| XM_005593332.2 | XP_005593389.1 | 3 | 1778 | 91 | ||
| XM_005593333.2 | XP_005593390.1 | 3 | 1775 | 91 | ||
| XM_015443342.1 | XP_015298828.1 | 4 | 1937 | 91 | ||
| GenBank mRNA | AB159441.1 | - | 1 | 1015 | 91 | |
| AB170630.1 | - | 3 | 1211 | 91 | ||
More Information
Set Membership: |
|

GPR34
Genomic Map