Assay Details
| Gene Symbol: PAQR8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005552754.2 | XP_005552811.1 | 2 | 1006 | 80 | |
| GenBank mRNA | AB174227.1 | - | 1 | 815 | 80 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

PAQR8| Gene Symbol: PAQR8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005552754.2 | XP_005552811.1 | 2 | 1006 | 80 | |
| GenBank mRNA | AB174227.1 | - | 1 | 815 | 80 | |
Set Membership: |
|