Assay Details
| Gene Symbol: SPTSSB | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015445272.1 | XP_015300758.1 | 1 - 2 | 1514 | 68 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

SPTSSB| Gene Symbol: SPTSSB | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015445272.1 | XP_015300758.1 | 1 - 2 | 1514 | 68 | |
Set Membership: |
|