Assay Details
| Gene Symbol: PCDHB12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015452021.1 | XP_015307507.1 | 1 | 3024 | 82 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_015452021.1 transcripts.
Set Membership: |
|

PCDHB12
Genomic Map