Assay Details
| Gene Symbol: SOCS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005591262.2 | XP_005591319.1 | 1 - 2 | 198 | 156 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_005591262.2 transcripts.
Set Membership: |
|

SOCS1
Genomic Map