Assay Details
| Gene Symbol: GPR45 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015433331.1 | XP_015288817.1 | 1 | 717 | 92 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_015433331.1 transcripts.
Set Membership: |
|

GPR45
Genomic Map