Assay Details
| Gene Symbol: LOC102121904 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005555157.1 | XP_005555214.1 | 6 | 2208 | 79 | |
| XM_005555160.1 | XP_005555217.1 | 5 | 1988 | 79 | ||
More Information
Set Membership: |
|

LOC102121904
Genomic Map