Assay Details
| Gene Symbol: Pten | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008960.2 | NP_032986.1 | 1 - 2 | 945 | 114 | |
| GenBank mRNA | AK076980.1 | - | 1 - 2 | 903 | 114 | |
| AK088717.1 | - | 1 - 2 | 924 | 114 | ||
| AK148736.1 | - | 1 - 2 | 783 | 114 | ||
| AK164302.1 | - | 1 - 2 | 928 | 114 | ||
| AK208703.1 | - | 1 - 2 | 177 | 114 | ||
| BC021445.1 | - | 1 - 2 | 294 | 5038272 | 114 | |
| U92437.1 | - | 1 - 2 | 1025 | 114 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Pten
Genomic Map