Assay Details
| Gene Symbol: Chrna7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_007390.3 | NP_031416.3 | 6 - 7 | 650 | 88 | |
| GenBank mRNA | AF225980.1 | - | 6 - 7 | 640 | 88 | |
| AK157809.1 | - | 6 - 7 | 647 | 88 | ||
| AY574266.1 | - | 6 - 7 | 600 | 88 | ||
| L37663.1 | - | 6 - 7 | 650 | 88 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Chrna7
Genomic Map