| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR1600012206.mm.1 |
NM_007470.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1600012206.mm.1 |
TC1600001479.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1600012209.mm.1 |
NM_007470.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1600012209.mm.1 |
TC1600001479.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1600012205.mm.1 |
NM_007470.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1600012205.mm.1 |
TC1600001479.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1600012216.mm.1 |
NM_007470.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1600012216.mm.1 |
TC1600001479.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1600012219.mm.1 |
NM_007470.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1600012219.mm.1 |
TC1600001479.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1600012204.mm.1 |
NM_007470.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1600012204.mm.1 |
TC1600001479.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1600012208.mm.1 |
NM_007470.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1600012208.mm.1 |
TC1600001479.mm.1 |
MTA TC |