Assay Details
| Gene Symbol: Atp6v1b2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_007509.3 | NP_031535.2 | 1 - 2 | 188 | 61 | |
| GenBank mRNA | AK146499.1 | - | 1 - 2 | 153 | 61 | |
| AK151200.1 | - | 1 - 2 | 189 | 61 | ||
| AK151322.1 | - | 1 - 2 | 161 | 61 | ||
| AK151586.1 | - | 1 - 2 | 161 | 61 | ||
| AK152718.1 | - | 1 - 2 | 161 | 61 | ||
| AK152766.1 | - | 1 - 2 | 161 | 61 | ||
| AK153341.1 | - | 1 - 2 | 153 | 61 | ||
| AK159133.1 | - | 1 - 2 | 159 | 61 | ||
| AK159153.1 | - | 1 - 2 | 159 | 61 | ||
| AK159586.1 | - | 1 - 2 | 156 | 61 | ||
| AK159701.1 | - | 1 - 2 | 160 | 61 | ||
| AK159986.1 | - | 1 - 2 | 160 | 61 | ||
| AK160080.1 | - | 1 - 2 | 159 | 61 | ||
| AK160854.1 | - | 1 - 2 | 154 | 61 | ||
| AK166669.1 | - | 1 - 2 | 128 | 61 | ||
| AK168852.1 | - | 1 - 2 | 152 | 61 | ||
| AK169155.1 | - | 1 - 2 | 167 | 61 | ||
| AK169270.1 | - | 1 - 2 | 164 | 61 | ||
| AK169420.1 | - | 1 - 2 | 156 | 61 | ||
| AK170110.1 | - | 1 - 2 | 184 | 61 | ||
| BC012497.1 | - | 1 - 2 | 153 | 4500843 | 61 | |
| BC046302.1 | - | 1 - 2 | 143 | 6492612 | 61 | |
| BC085300.1 | - | 1 - 2 | 135 | 6305977 | 61 | |
| Y12634.1 | - | 1 - 2 | 145 | 61 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Atp6v1b2
Genomic Map