Assay Details
| Gene Symbol: Atp6v1b2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_007509.3 | NP_031535.2 | 3 - 4 | 348 | 78 | |
| GenBank mRNA | AK146499.1 | - | 3 - 4 | 313 | 78 | |
| AK151200.1 | - | 3 - 4 | 349 | 78 | ||
| AK151322.1 | - | 3 - 4 | 321 | 78 | ||
| AK151586.1 | - | 3 - 4 | 321 | 78 | ||
| AK152718.1 | - | 3 - 4 | 321 | 78 | ||
| AK152766.1 | - | 3 - 4 | 321 | 78 | ||
| AK153341.1 | - | 3 - 4 | 313 | 78 | ||
| AK159133.1 | - | 3 - 4 | 319 | 78 | ||
| AK159153.1 | - | 3 - 4 | 319 | 78 | ||
| AK159586.1 | - | 3 - 4 | 316 | 78 | ||
| AK159701.1 | - | 3 - 4 | 320 | 78 | ||
| AK159986.1 | - | 3 - 4 | 320 | 78 | ||
| AK160080.1 | - | 3 - 4 | 319 | 78 | ||
| AK160854.1 | - | 3 - 4 | 314 | 78 | ||
| AK166669.1 | - | 3 - 4 | 288 | 78 | ||
| AK168852.1 | - | 3 - 4 | 312 | 78 | ||
| AK169155.1 | - | 3 - 4 | 327 | 78 | ||
| AK169270.1 | - | 3 - 4 | 324 | 78 | ||
| AK169420.1 | - | 3 - 4 | 316 | 78 | ||
| AK170110.1 | - | 3 - 4 | 344 | 78 | ||
| AK182828.1 | - | 3 - 4 | 203 | 78 | ||
| AK185732.1 | - | 3 - 4 | 172 | 78 | ||
| AK205457.1 | - | 3 - 4 | 183 | 78 | ||
| AK206476.1 | - | 3 - 4 | 183 | 78 | ||
| AK208817.1 | - | 2 - 3 | 150 | 78 | ||
| AK209288.1 | - | 3 - 4 | 182 | 78 | ||
| AK209472.1 | - | 3 - 4 | 183 | 78 | ||
| AK213950.1 | - | 3 - 4 | 183 | 78 | ||
| AK214894.1 | - | 3 - 4 | 179 | 78 | ||
| AK215019.1 | - | 3 - 4 | 181 | 78 | ||
| AK216107.1 | - | 2 - 3 | 145 | 78 | ||
| BC012497.1 | - | 3 - 4 | 313 | 4500843 | 78 | |
| BC046302.1 | - | 3 - 4 | 303 | 6492612 | 78 | |
| BC085300.1 | - | 3 - 4 | 295 | 6305977 | 78 | |
| Y12634.1 | - | 3 - 4 | 305 | 78 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Atp6v1b2
Genomic Map