Assay Details
| Gene Symbol: Atp6v1b2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_007509.3 | NP_031535.2 | 4 - 5 | 440 | 62 | |
| GenBank mRNA | AK146499.1 | - | 4 - 5 | 405 | 62 | |
| AK151200.1 | - | 4 - 5 | 441 | 62 | ||
| AK151322.1 | - | 4 - 5 | 413 | 62 | ||
| AK151586.1 | - | 4 - 5 | 413 | 62 | ||
| AK152718.1 | - | 4 - 5 | 413 | 62 | ||
| AK152766.1 | - | 4 - 5 | 413 | 62 | ||
| AK159133.1 | - | 4 - 5 | 411 | 62 | ||
| AK159153.1 | - | 4 - 5 | 411 | 62 | ||
| AK159586.1 | - | 4 - 5 | 408 | 62 | ||
| AK159701.1 | - | 4 - 5 | 412 | 62 | ||
| AK159986.1 | - | 4 - 5 | 412 | 62 | ||
| AK160080.1 | - | 4 - 5 | 411 | 62 | ||
| AK160854.1 | - | 4 - 5 | 406 | 62 | ||
| AK166669.1 | - | 4 - 5 | 380 | 62 | ||
| AK168852.1 | - | 4 - 5 | 404 | 62 | ||
| AK169155.1 | - | 4 - 5 | 419 | 62 | ||
| AK169270.1 | - | 4 - 5 | 416 | 62 | ||
| AK169420.1 | - | 4 - 5 | 408 | 62 | ||
| AK170110.1 | - | 4 - 5 | 436 | 62 | ||
| AK191015.1 | - | 3 - 4 | 258 | 62 | ||
| AK205457.1 | - | 4 - 5 | 275 | 62 | ||
| AK206476.1 | - | 4 - 5 | 275 | 62 | ||
| AK206807.1 | - | 3 - 4 | 237 | 62 | ||
| AK207516.1 | - | 3 - 4 | 237 | 62 | ||
| AK209288.1 | - | 4 - 5 | 274 | 62 | ||
| AK209472.1 | - | 4 - 5 | 275 | 62 | ||
| AK209631.1 | - | 4 - 5 | 275 | 62 | ||
| AK212223.1 | - | 4 - 5 | 275 | 62 | ||
| AK213950.1 | - | 4 - 5 | 275 | 62 | ||
| AK214894.1 | - | 4 - 5 | 271 | 62 | ||
| AK215019.1 | - | 4 - 5 | 273 | 62 | ||
| AK215413.1 | - | 3 - 4 | 239 | 62 | ||
| AK216107.1 | - | 3 - 4 | 237 | 62 | ||
| AK217254.1 | - | 4 - 5 | 276 | 62 | ||
| BC012497.1 | - | 4 - 5 | 405 | 4500843 | 62 | |
| BC046302.1 | - | 4 - 5 | 395 | 6492612 | 62 | |
| BC085300.1 | - | 4 - 5 | 387 | 6305977 | 62 | |
| Y12634.1 | - | 4 - 5 | 397 | 62 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Atp6v1b2
Genomic Map